Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGD 94-1, Tocris Bioscience™

Catalog Number 329850
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
178928-68-2
C18H20N6
320.4
OTVQCHUIPJYASM-UHFFFAOYSA-N
unii-do9m06bfrj, do9m06bfrj, 2-4-2-phenyl-1h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, 2-4-2-phenyl-1h-imidazol-5-yl methyl piperazin-1-yl pyrimidine, 2-phenyl-4 5-4-2-pyrimidinyl-piperazin-1-yl methyl imidazole dimaleate, 2-4-2-phenyl-1h-imidazol-5-yl methyl-1-piperazinyl-pyrimidine, 2-4-2-phenyl-3h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, d0f7nm, d0l3ji
188942
2-[4-[(2-phenyl-1H-imidazol-5-yl)methyl]piperazin-1-yl]pyrimidine
C1CN(CCN1CC2=CN=C(N2)C3=CC=CC=C3)C4=NC=CC=N4
This item is not returnable. View return policy
178928-68-2
C18H20N6
320.4
OTVQCHUIPJYASM-UHFFFAOYSA-N
unii-do9m06bfrj, do9m06bfrj, 2-4-2-phenyl-1h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, 2-4-2-phenyl-1h-imidazol-5-yl methyl piperazin-1-yl pyrimidine, 2-phenyl-4 5-4-2-pyrimidinyl-piperazin-1-yl methyl imidazole dimaleate, 2-4-2-phenyl-1h-imidazol-5-yl methyl-1-piperazinyl-pyrimidine, 2-4-2-phenyl-3h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, d0f7nm, d0l3ji
188942
2-[4-[(2-phenyl-1H-imidazol-5-yl)methyl]piperazin-1-yl]pyrimidine
C1CN(CCN1CC2=CN=C(N2)C3=CC=CC=C3)C4=NC=CC=N4

Selective D4 receptor ligand

High affinity D4 receptor ligand; selective over D1, D2, D3 and D5 receptors. Displays antagonist activity at the human D4.2 receptor (Ki = 3.6 nM in transfected CHO cells) and exhibits agonist activity at the D4.4 receptor in HEK 293 cells.
Quantity 50 mg
Formula Weight 320.39
Percent Purity >99%
Chemical Name or Material NGD 94-1
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit