Learn More
NGDN Mouse anti-Human, Clone: OTI1B8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808822
Catalog No. 50-166-9103
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
Specifications
Specifications
NGDN
Monoclonal
Unconjugated
NGDN
Lyophilized
NGDN
Full length human recombit protein of human NGDN produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1B8
PBS with 8% trehalose and no preservative; pH 7.3
Q8NEJ9
C14orf120, CANu1, LCP5, NGD, lpd-2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
25983