Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NGDN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587151
Description
Purity is > 95% by SDS-PAGE.
Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
Specifications
NGDN | |
Polyclonal | |
Unconjugated | |
Ngdn | |
1500001L15Rik; C14orf120; CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; LCP5; lpd-2; neuroguidin; neuroguidin, EIF4E binding protein; NGD; Ngdn; RGD1310211 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25983, 305887, 68966 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NEJ9, Q9DB96 | |
Ngdn | |
Recombinant fusion protein of human NGDN. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction