Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGDN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548723

Catalog No. PA548723

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
Specifications

Specifications

NGDN
Polyclonal
PBS with 0.09% sodium azide
Q8NEJ9
Neuroguidin, Centromere accumulated nuclear protein 1, CANu1, EIF4E-binding protein, NGDN, C14orf120
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25983
Western Blot
Unconjugated
NGDN
Liquid
NGDN
KLH conjugated synthetic peptide between 136-165 amino acids from the Central region of human NGDN.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit