Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGDN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 165241AP

Catalog No. 50-173-1007


Add to cart

Description

Description

Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
Specifications

Specifications

NGDN
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NEJ9, Q9DB96
C14orf120, EIF4E binding protein, LCP5, lpd 2, Neuroguidin, NGD, NGDN
Rabbit
IgG
150 μL
-20°C
Primary
25983, 305887, 68966
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NGDN
Liquid
Ngdn
NGDN Fusion Protein Ag9763
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit