Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NGF Monoclonal Antibody (C2)
Mouse Monoclonal Antibody
$184.40 - $493.00
Specifications
Antigen | NGF |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541645
|
Invitrogen™
MA541645 |
200 μL |
Each for $493.00
|
|
|||||
PIMA543759
|
Invitrogen™
MA543759 |
20 μL |
Each for $184.40
|
|
|||||
Description
NGF Monoclonal Antibody for Western Blot
Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.Specifications
NGF | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P01138 | |
4803 | |
Recombinant protein Nerve Growth Factor. The antigen corresponds to amino acid range 19-239 of the target protein. | |
Antibody |
C2 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
NGF | |
B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF ß; Ngfb; NGFbeta; NGFß; ß nerve growth factor; ß NGF; ßNGF | |
NGF | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title