Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGFI-Balpha/Nur77/NR4A1 Rabbit anti-Human, Clone: JM59-11, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266980
Description
NGFI-Balpha/Nur77/NR4A1 Monoclonal antibody specifically detects NGFI-Balpha/Nur77/NR4A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NGFI-Balpha/Nur77/NR4A1 | |
JM59-11 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40% Glycerol with 0.05% Sodium Azide | |
Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin | |
Purified | |
NR4A1 | |
Recombinant protein | |
Protein A purified | |
Apoptosis | |
Primary | |
3164 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title