Learn More
NGFR Mouse anti-Human, Clone: SPM299, Abnova™
Mouse monoclonal antibody raised against native human NGFR.
Manufacturer:  Abnova Corporation MAB13130
Catalog No. 89-934-698
Add to cart
Description
Description
Nerve growth factor receptor contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain. [provided by RefSeq]
Specifications
Specifications
NGFR
SPM299
Mouse monoclonal antibody raised against native human NGFR.
In 10mM PBS.
CD271/Gp80-LNGFR/TNFRSF16/p75(NTR)/p75NTR
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
NGFR
A875 melanoma cells.
Protein A/G purification
RUO
Primary
4804