Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGFR Rabbit anti-Human, Unconjugated, Clone: NGFR/1997R, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation RAB00594
Description
NGFR Monoclonal antibody specifically detects NGFR in Human, Non-human Primate samples. It is validated for IHC-P.Specifications
NGFR | |
Monoclonal | |
Unconjugated | |
NGFR | |
Rabbit | |
100 μg | |
4804 | |
Store at -20 to -80°C | |
IgG |
Immunohistochemistry (Paraffin) | |
NGFR/1997R | |
nerve growth factor receptor (TNFR superfamily, member 16) | |
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR | |
Recombinant protein corresponding to full-length human NGFR | |
Primary | |
Human, Non-human Primate | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction