Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFR/TNFRSF16/p75NTR Rabbit anti-Human, Clone: NGFR/2550R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen NGFR/TNFRSF16/p75NTR
Clone NGFR/2550R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122290
View Documents
Novus Biologicals
NBP30740820UG
20 μg
Each for $159.82
Add to cart
 
NB122289
View Documents
Novus Biologicals
NBP30740810UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

NGFR/TNFRSF16/p75NTR Monoclonal antibody specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

NGFR/TNFRSF16/p75NTR
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
Human
CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16
Recombinant human NGFR/TNFRSF16/p75NTR protein fragment (around aa 281-421) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
NGFR/2550R
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Apoptosis, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
10 mM PBS with 0.05% BSA
4804
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit