Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NgR2/NgRH1/RTN4RL2 Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 532, Clone: P14, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252676AF532

Catalog No. NP22676A32


Add to cart

Description

Description

NgR2/NgRH1/RTN4RL2 Monoclonal antibody specifically detects NgR2/NgRH1/RTN4RL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

NgR2/NgRH1/RTN4RL2
P14
Western Blot, Flow Cytometry, ELISA
Conjugated, Purified
RTN4RL2
(His)6-tagged fragment of rat NgR2 (amino acids 279-420).
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
The antibody binds to human NgR2 with a Kd of 2.3 nM and to rat NgR2 with a Kd of 0.48 nM (bivalent binding).
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
NgR2, NGRH1, reticulon 4 receptor-like 2
Rabbit
IgG Kappa
0.1 mL
Primary
349667.0
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit