Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 750, Clone: P14, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252676AF750
Description
NgR2/NgRH1/RTN4RL2 Monoclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
NgR2/NgRH1/RTN4RL2 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
RTN4RL2 | |
(His)6-tagged fragment of rat NgR2 (amino acids 279-420). | |
Primary | |
The antibody binds to human NgR2 with a Kd of 2.3 nM and to rat NgR2 with a Kd of 0.48 nM (bivalent binding). | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA | |
P14 | |
Western Blot, Flow Cytometry, ELISA | |
NgR2, NGRH1, reticulon 4 receptor-like 2 | |
Rabbit | |
0.1 mL | |
349667 | |
Human, Mouse, Rat | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title