Learn More
Medchemexpress LLC NH2-C2-NH-Boc (PROTAC Linker 22) | 57260-73-8 | 99.9% | 160.21 | 1 G
SDP
Supplier:  Medchemexpress LLC HY401711G
Encompass_Preferred
NH2-C2-NH-Boc (PROTAC Linker 22) is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are molecules containing two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and is not sold to patients.
  • Utilized in the synthesis of PROTACs.
  • Contains two different ligands connected by a linker.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • For research use only.
Catalog No. 50-004-03816