Learn More
Medchemexpress LLC NH2-C4-NH-Boc | 68076-36-8 | 99.98% | 188.27 | 10 G
SDP
Supplier:  Medchemexpress LLC HY4017810G
Encompass_Preferred
NH2-C4-NH-Boc is a PROTAC linker, characterized by its alkyl/ether composition. It is utilized in the synthesis of various proteolysis targeting chimeras (PROTACs). PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins, containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • Alkyl/ether composition
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-03947