Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$487.50
Specifications
Antigen | NHE8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NHE8 Polyclonal specifically detects NHE8 in Human, Mouse samples. It is validated for Western Blot.Specifications
NHE8 | |
Polyclonal | |
Rabbit | |
Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism | |
DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8 | |
SLC9A8 | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Q2M1U9 | |
23315 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title