Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHEDC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548902

Catalog No. PA548902

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Electroneutral exchange of protons for Na(+) and Li(+) across the inner mitochondrial membrane. Contributes to the organellar volume homeostasis. Required for osteoclast differentiation and bone resorption activity.
Specifications

Specifications

NHEDC2
Polyclonal
PBS with 0.09% sodium azide
Q86UD5
Mitochondrial sodium/hydrogen exchanger 9B2, Mitochondrial Na(+)/H(+) exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, NHE domain-containing protein 2, Sodium/hydrogen exchanger-like domain-containing protein 2, Solute carrier family 9 subfamily B member 2, SLC9B2, NHA2, NHEDC2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
133308
Western Blot
Unconjugated
SLC9B2
Liquid
SLC9B2
KLH conjugated synthetic peptide between 458-486 amino acids from the C-terminal region of human NHEDC2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit