Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHEJ1 Mouse anti-Human, Clone: OTI1G5, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502176
Description
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.Specifications
NHEJ1 | |
Monoclonal | |
1.6 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H9Q4 | |
XLF | |
Mouse | |
IgG2b | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
79840 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G5 | |
Unconjugated | |
NHEJ1 | |
Liquid | |
NHEJ1 | |
Full length human recombit protein of human NHEJ1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title