Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NHI 2
LDHA inhibitor
$302.76 - $1222.64
Specifications
Purity | >98% |
---|---|
Description | NHI 2 |
Molecular Weight (g/mol) | 335.28 |
Product Type | NHI 2 |
Description
LDHA inhibitor; reduces lactate production in HeLa cells. Inhibits cell growth of LPC006 and PANC-1 cells in vitro. Enhances the antiproliferative effects of Gemcitabine (Cat. No. 3259) on LPC006 and PANC-1 cells. Also inhibits ferroptosis.Specifications
>98% | |
NHI 2 | |
335.28 | |
NHI 2 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title