Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHLRC1 Mouse anti-Human, Biotin, Clone: S85-18, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242173B

Catalog No. NBP242173B


Add to cart

Description

Description

NHLRC1 Monoclonal antibody specifically detects NHLRC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NHLRC1
S85-18
Western Blot, Immunocytochemistry/Immunofluorescence
Q6VVB1, NP_940988.2
bA204B7.2, E3 ubiquitin-protein ligase NHLRC1, EC 6.3.2.-, EPM2A, EPM2BMGC119264, MALIN, MGC119262, MGC119265, NHL repeat containing 1, NHL repeat-containing protein 1
Mouse
IgG1
0.1 ml
Primary
378884
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
Purified
NHLRC1
Fusion protein amino acids 2-125 (N-terminus encompassing RING domain) of human Malin. 86% identical to rat, and 77% identical to mouse.
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 42kDa.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit