Learn More
NHLRC1 Mouse anti-Human, DyLight 405, Clone: S85-18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242173V
Catalog No. NBP242173V
Add to cart
Description
Description
NHLRC1 Monoclonal antibody specifically detects NHLRC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
NHLRC1
S85-18
Western Blot, Immunocytochemistry/Immunofluorescence
Q6VVB1, NP_940988.2
bA204B7.2, E3 ubiquitin-protein ligase NHLRC1, EC 6.3.2.-, EPM2A, EPM2BMGC119264, MALIN, MGC119262, MGC119265, NHL repeat containing 1, NHL repeat-containing protein 1
Mouse
IgG1
0.1 ml
Primary
378884
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
NHLRC1
Fusion protein amino acids 2-125 (N-terminus encompassing RING domain) of human Malin. 86% identical to rat, and 77% identical to mouse.
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 42kDa.