Learn More
NHLRC1 Mouse anti-Human, HRP, Clone: S85-18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242173H
Catalog No. NBP242173H
Add to cart
Description
Description
NHLRC1 Monoclonal antibody specifically detects NHLRC1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
NHLRC1
S85-18
Western Blot
Q6VVB1, NP_940988.2
bA204B7.2, E3 ubiquitin-protein ligase NHLRC1, EC 6.3.2.-, EPM2A, EPM2BMGC119264, MALIN, MGC119262, MGC119265, NHL repeat containing 1, NHL repeat-containing protein 1
Mouse
IgG1
0.1 ml
Primary
378884
Human
Western Blot
HRP
PBS with No Preservative
Purified
NHLRC1
Fusion protein amino acids 2-125 (N-terminus encompassing RING domain) of human Malin. 86% identical to rat, and 77% identical to mouse.
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 42kDa.