Learn More
NHP2L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158021AP
Catalog No. 50-173-0490
Add to cart
Description
Description
Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5 ft. stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
NHP2L1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P55769, Q9D0T1
15.5K, FA 1, FA1, hSNU13, NHP2 like protein 1, NHP2L1, NHPX, OTK27, SNRNP15 5, SNU13, SNU13 homolog, SPAG12, SSFA1
Rabbit
IgG
150 μL
-20°C
Primary
20826, 4809
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SNU13
Liquid
SNU13
NHP2L1 Fusion Protein Ag8532
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse