Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NI 42

High affinity BRPF inhibitor

$189.00 - $795.00

Specifications

Chemical Name or Material 4-Cyano-N-(1,2-dihydro-1,3-dimethyl-2-oxo-6-quinolinyl)benzenesulfonamide
Molecular Formula C18H15N3O3S
CAS 1884640-99-6
Product Type Target: Bromodomain Inhibitors
Formula Weight 353.39
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-861-0 View Documents Tocris Bioscience™
5786/10
10mg Each for $189.00
Add to cart
 
57-865-0 View Documents Tocris Bioscience™
5786/50
50mg Each for $795.00
Add to cart
 
Description

Description

High affinity BRPF inhibitor (Kd values are 40, 210 and 943 nM for BRPF1B, 2 and 3, respectively). Also exhibits modest activity at BRD9 (Kd = 1130 nM). Exhibits selectivity for BRPF over 44 other bromodomains. Orally bioavailable.
Specifications

Specifications

4-Cyano-N-(1,2-dihydro-1,3-dimethyl-2-oxo-6-quinolinyl)benzenesulfonamide
1884640-99-6
353.39
C18H15N3O3S
Target: Bromodomain Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit