Learn More
GE Healthcare Ni Sepharose™ 6 Fast Flow: PROMO
Product
Manufacturer:  GE Healthcare 17531802PM
Catalog No. 45-002-985LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Ni Sepharose 6 Fast Flow is a member of the GE Healthcare range of IMAC chromatography media for capture and intermediate purification.
  • Designed for scaling up purification of histidine-tagged proteins.
  • High protein binding capacity and minimal leakage of Ni2+ ions.
  • As a BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
  • Available in convenient prepacked HisPrep and HisTrap column formats.
  • Composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2+ ions for purification of histidine tagged proteins.
  • The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2+ leakage while providing high capacity.
  • Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
  • As member of the BioProcess media range, Ni Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Specifications
Specifications
0.1M HCl, 0.1M NaOH, 8M Urea
100mL
250mm
1 bar
90μm
4° to 30°C
Ni Sepharose 6 FF chromato graphy media
9.84 in.
600cm/hr.
3 to 12
6% Cross-linked Agarose