Learn More
GE Healthcare Ni Sepharose™ High Performance Media PROMO
Precharged with Ni2+ for optimal purification of histidine-tagged proteins.  GE Healthcare Lifescience™ Ni Sepharose™ High Performance Media shows high binding capacity and compatibility.
Manufacturer:  GE Healthcare 29361659
Catalog No. 45-000-329PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Negligible leakage of Ni2+ ions
  • High binding capacity: at least 40mg protein/mL medium
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Stable (long-term) in the pH range 3-12
  • Average particle size: 34µm
Specifications
Specifications
Ni Sepharose HP chromato graphy media
3 to 12
6% Cross-linked Agarose
100mL
34μm
4° to 30°C