Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niacin, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
25g
500g
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
59-67-6
C6H5NO2
123.111
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid,niacin,3-pyridinecarboxylic acid,3-carboxypyridine,wampocap,acidum nicotinicum,apelagrin,pellagrin,akotin,daskil
938
CHEBI:15940
pyridine-3-carboxylic acid
C1=CC(=CN=C1)C(=O)O
59-67-6
C6H5NO2
123.111
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid,niacin,3-pyridinecarboxylic acid,3-carboxypyridine,wampocap,acidum nicotinicum,apelagrin,pellagrin,akotin,daskil
938
CHEBI:15940
pyridine-3-carboxylic acid
C1=CC(=CN=C1)C(=O)O
Specific Gravity 1L = 1.47kg
Quantity 500g
Packaging Wide Mouth PVC Coated Glass Bottle
Formula Weight 123.11
Grade FCC/USP
Chemical Name or Material Niacin