Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niacin, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
25g
500g
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle

Chemical Identifiers

59-67-6
C6H5NO2
123.111
PVNIIMVLHYAWGP-UHFFFAOYSA-N
938
CHEBI:15940
Specific Gravity 1L = 1.47kg
Quantity 25g
Packaging Wide Mouth Amber Glass Bottle
SMILES C1=CC(=CN=C1)C(=O)O
IUPAC Name pyridine-3-carboxylic acid
Formula Weight 123.11
Chemical Name or Material Niacin
Grade FCC/USP