Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niacinamide, 98.5 to 101.5% (Dry Basis), Affymetrix/USB™

Click to view available options
Quantity:
1kg
10kg
Packaging:
Plastic bottle
98-92-0
C6H6N2O
122.127
MFCD00006395
DFPAKSUCGFBDDF-UHFFFAOYSA-N
nicotinamide,niacinamide,3-pyridinecarboxamide,vitamin pp,nicotinic acid amide,papulex,aminicotin,amixicotyn,nicobion,nicotylamide
936
CHEBI:17154
pyridine-3-carboxamide
C1=CC(=CN=C1)C(=O)N
98-92-0
C6H6N2O
122.127
MFCD00006395
DFPAKSUCGFBDDF-UHFFFAOYSA-N
nicotinamide,niacinamide,3-pyridinecarboxamide,vitamin pp,nicotinic acid amide,papulex,aminicotin,amixicotyn,nicobion,nicotylamide
936
CHEBI:17154
pyridine-3-carboxamide
C1=CC(=CN=C1)C(=O)N
Packaging Plastic bottle
Quantity 10kg
Flash Point ≥110°C (230°F)
Specific Gravity 1.40
Melting Point 128°C to 132°C
Formula Weight 122.12
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Chemical Name or Material Niacinamide
Assay Dry basis