Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niban-like protein Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549675

Catalog No. PA549675


Add to cart

Description

Description

The antibody detects endogenous level of total Niban-like protein .

The Niban-like protein is encoded by a maternal imprinting gene. This protein may possess growth suppressor activity. Two major alternatively spliced isoforms have been described. forms. One form, Meg3-proximal (Meg3p), contains exons 1-3. The second form, Meg3-distal (Meg3d) did not contain exons 1-3 and was present in oocytes and in 1- and 2-cell embryos. Adachi H, Majima S, et al. Oncogene. 2004 Apr 22;23(19):3495-500 Matsumoto F, Fujii H, et al. Hum Pathol. 2006 Dec; 37(12):1592-600.
Specifications

Specifications

Niban-like protein
Polyclonal
Unconjugated
FAM129B
Liquid
FAM129B
Peptide sequence around aa.710∽714 (Q-V-S-S-P) derived from Human Niban-like protein.
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96TA1, Q8R1F1
C9orf88
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
227737, 64855
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit