Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicastrin Mouse, HRP, Clone: 3B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130175H

Catalog No. NBP130175H


Add to cart

Description

Description

Nicastrin Monoclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Nicastrin
3B2
Western Blot, ELISA
Purified
NCSTN
Recombinant human Nicastrin (34-153aa) purified from E. coli
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Nicastrin protein.
ELISA, Western Blot
HRP
PBS with No Preservative
anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin
Mouse
IgG2b Kappa
0.1 ml
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary
23385
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit