Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicastrin Rabbit anti-Human, Alexa Fluor 647, Clone: 4, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289826AF647

Catalog No. NB109584


Add to cart

Description

Description

Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Nicastrin
4
50 mM Sodium Borate
anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin
This antibody was obtained from a rabbit immunized with purified, recombinant Human Nicastrin (NP_056146.1; Met1-Glu669).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Immunofluorescence
Alexa Fluor 647
Purified
Rabbit
IgG
0.1 mL
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary
23385
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit