Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicergoline, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
27848-84-6
C24H26BrN3O3
484.394
YSEXMKHXIOCEJA-FVFQAYNVSA-N
nicergoline,nicotergoline,nimergoline,nimergoline base,nicergolinum,nicergolin,nicergolina,sermion,nicergolin german,nicergolina dcit
34040
[(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br
This item is not returnable. View return policy
27848-84-6
C24H26BrN3O3
484.394
YSEXMKHXIOCEJA-FVFQAYNVSA-N
nicergoline,nicotergoline,nimergoline,nimergoline base,nicergolinum,nicergolin,nicergolina,sermion,nicergolin german,nicergolina dcit
34040
[(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br

α antagonist

α-adrenergic, vasodilator. Cognitive enhancer.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 484.39
Percent Purity >98%
Chemical Name or Material Nicergoline

Recommended Storage : Store at Room Temperature