Learn More
Nichrome (Chrome) Wire Products for Science Education
Resists oxidation and really takes the heat.
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
S49085
View Documents
Sempco Inc
WS0103
N/A 22AWG This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Durable alloy wire available in various gauges; lighter gauges can be used for needles and inoculation loops.
  • Heats to 1093°C (2000°F)
  • Provides very efficient heating capacity
  • Highly resistant to oxidation
  • Supplied in 0.25 lb. spools