Nichrome etchant, Alfa Aesar™

High Purity Ceric Ammonium Nitrate System Provides Clean, Precise Etching of Nichrome Layers For Preparation of Thin Film Circuits

Manufacturer: Alfa Aesar 44585R0

Quantity 1 qt.

 View more versions of this product

Catalog No. AA44585R0

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    MDL Number MFCD07370976
    Chemical Name or Material Nichrome etchant

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 1 qt.
    DOT Information DOT Class: 8, Packaging Group: III
    Health Hazard 2 When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    Odor Acrid
    Vapor Pressure 17mm Hg at 20°C
    MDL Number MFCD07370976
    Health Hazard 1 H360-H362-H314-H290-H333: May damage fertility or the unborn child. May cause harm to breast-fed children. Causes severe skin burns and eye damage. May be corrosive to metals. May be harmful if inhaled.
    Chemical Name or Material Nichrome etchant

    When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H360-H362-H314-H290-H333: May damage fertility or the unborn child. May cause harm to breast-fed children. Causes severe skin burns and eye damage. May be corrosive to metals. May be harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit