Learn More
Nichrome etchant, Thermo Scientific Chemicals
High Purity Ceric Ammonium Nitrate System Provides Clean, Precise Etching of Nichrome Layers For Preparation of Thin Film Circuits
Supplier: Thermo Scientific Chemicals 044585LV
Description
High purity ceric ammonium nitrate system provides clean, precise etching of nichrome layers for preparation of thin film circuits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless | |
1.25g/cm3 at 20°C | |
1 gal. | |
17mm Hg at 20°C |
Nichrome etchant | |
MFCD07370976 | |
Acrid | |
Liquid |
Safety and Handling
When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P201-P202-P234-P260-P263-P264b-P270-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H331-H360D-H362
DOTInformation : DOT Class: 8, Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only