Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel, 99.9%, (trace metal basis), powder, -325 mesh, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
7440-02-0
Ni
58.693
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
7440-02-0
Ni
58.693
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
For ultra trace analysis
Quantity 100g
CAS Min % 99.85
CAS Max % 100.0
Density 8.9
Assay Percent Range 99.9%
Fieser 12,335; 13,197; 17,296
Merck Index 15, 6583
Packaging Plastic bottle
Formula Weight 58.7
Physical Form Fine Powder
Percent Purity 99.9%
Grade Trace Metal Basis
Total Trace Metal Impurities 0.1% max.
Color Brown-Gray
Melting Point 1455.0°C
Boiling Point 2730.0°C
Chemical Name or Material Nickel, 99.9%, -325 mesh
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 231-111-4

RTECSNumber : QR5950000

TSCA : TSCA