Alfa Aesar™ Nickel Aluminum, Raney™ type non-activated

Manufacturer: Alfa Aesar™ 8767618

Quantity 50g

 View more versions of this product

Catalog No. AA8767618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12635-29-9
    MDL Number MFCD00167392
    UN Number UN3208
    Synonym Aluminum Nickel, Raney™ type; Aluminum Nickel
    Chemical Name or Material Nickel Aluminum, Raney™ type non-activated

    Specifications

    Synonym Aluminum Nickel, Raney™ type; Aluminum Nickel
    Health Hazard 2 GHS H Statement
    H228-H261-H372-H351-H317
    Flammable solid.
    In contact with water releases flammable gas.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P260-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 50g
    Physical Form Powder
    MDL Number MFCD00167392
    UN Number UN3208
    Health Hazard 1 Danger
    Color Gray to black
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
    Avg. Mol. Wt. or Mol. Wt. Range Ni:Al
    CAS 12635-29-9
    Odor Odorless
    Chemical Name or Material Nickel Aluminum, Raney™ type non-activated

    GHS H Statement
    H228-H261-H372-H351-H317
    Flammable solid.
    In contact with water releases flammable gas.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P231+P232-P260-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit