Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel antimonide, 99.5% (metals basis excluding Co), Co 0.1-1%

Manufacturer:  Alfa Aesar™ 3626718

Quantity 50g

 View more versions of this product

Catalog No. AA3626718


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12035-52-8
MFCD00050100
84272
180.46
UN1549
Nickel antimonide

Specifications

12035-52-8
180.46
UN1549
10mm & down
Nickel antimonide
Lumps
MFCD00050100
84272
50g
99.5%, (metals basis excluding Co), Co 0.1 to 1%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H334-H351-H302-H317
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit