Alfa Aesar™ Nickel antimonide, 99.5% (metals basis excluding Co), Co 0.1-1%

Manufacturer: Alfa Aesar™ 3626709

Quantity 10g

 View more versions of this product

Catalog No. AA3626709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12035-52-8
    Formula Weight 180.46
    MDL Number MFCD00050100
    UN Number UN1549
    Beilstein 84272
    Chemical Name or Material Nickel antimonide

    Specifications

    Physical Form Lumps
    Formula Weight 180.46
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00050100
    UN Number UN1549
    Beilstein 84272
    Quantity 10g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
    TSCA TSCA
    Particle Size 10mm & down
    CAS 12035-52-8
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H334-H351-H302-H317
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Chemical Name or Material Nickel antimonide
    Assay 99.5%, (metals basis excluding Co), Co 0.1 to 1%

    GHS H Statement
    H331-H334-H351-H302-H317
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit