Alfa Aesar™ Nickel arsenide, 99.5% (metals basis excluding Co), Co 0.1-1%

Manufacturer: Alfa Aesar™ 3626914

Quantity 25g

 View more versions of this product

Catalog No. AA3626914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12068-61-0
    MDL Number MFCD00151450
    UN Number UN3077
    Chemical Name or Material Nickel arsenide

    Specifications

    Physical Form Lumps
    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Odor Odorless
    UN Number UN3077
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Particle Size 10mm & down
    MDL Number MFCD00151450
    CAS 12068-61-0
    Quantity 25g
    Chemical Name or Material Nickel arsenide
    Assay 99.5%, (metals basis excluding Co), Co 0.1 to 1%

    GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit