Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel arsenide, 99.5% (metals basis excluding Co), Co 0.1-1%

Manufacturer:  Alfa Aesar™ 3626906

Quantity 5g

 View more versions of this product

Catalog No. AA3626906


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12068-61-0
MFCD00151450
UN3077
Nickel arsenide

Specifications

12068-61-0
MFCD00151450
Odorless
5g
99.5%, (metals basis excluding Co), Co 0.1 to 1%
Lumps
UN3077
10mm & down
Nickel arsenide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit