Alfa Aesar™ Nickel based steam reforming catalyst, HiFUEL™ R110

Manufacturer: Alfa Aesar™ 4546536

Quantity 500g

 View more versions of this product

Catalog No. AA4546536

  Check Availability
Add to Cart

    Chemical Identifiers

    Chemical Name or Material Nickel based steam reforming catalyst HiFUEL™ R110

    Specifications

    Physical Form 4-Hole, 4-Flute Domed Cylinders
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 500g
    Health Hazard 1 Danger
    TSCA TSCA
    Color Pale gray to green
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Chemical Name or Material Nickel based steam reforming catalyst HiFUEL™ R110

    GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit