Nickel boride, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 1310914

Quantity 25g

 View more versions of this product

Catalog No. AA1310914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12007-01-1
    Physical Form -35 Mesh Granular
    Assay Percent Range 99%
    Molecular Formula Ni2B
    Formula Weight 128.23
    MDL Number MFCD00084757
    Chemical Name or Material Nickel boride

    Specifications

    Formula Weight 128.23
    Quantity 25g
    EINECS Number 234-494-6
    Physical Form -35 Mesh Granular
    Assay Percent Range 99%
    Molecular Formula Ni2B
    MDL Number MFCD00084757
    CAS 12007-01-1
    Chemical Name or Material Nickel boride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit