Alfa Aesar™ Nickel Chromium Aluminum Copper wire, 0.025mm (0.001 in.) dia., annealed (Evanohm™)

Manufacturer: Alfa Aesar™ 43635G9

Quantity 25m

 View more versions of this product

Catalog No. AA43635G9

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02684522
    Chemical Name or Material Nickel Chromium Aluminum Copper wire, 0.025mm (0.001 in.) dia., Annealed (Evanohm™)

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Wire
    Health Hazard 2 GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr:Al:Cu; 75:20:2.5:2.5 wt%
    Weight ≈0.0041g/m
    MDL Number MFCD02684522
    Health Hazard 1 Warning
    Quantity 25m
    Chemical Name or Material Nickel Chromium Aluminum Copper wire, 0.025mm (0.001 in.) dia., Annealed (Evanohm™)

    GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit