Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Chromium gauze, Alfa Aesar™

Open area: 36%; Width of opening: 0.015 in.

Manufacturer:  Alfa Aesar 40941FL

Quantity 75 x 75mm

 View more versions of this product

Catalog No. AA40941FL


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

11106-97-1
Ni:Cr; 80:20 wt%
Nickel Chromium, gauze
Wire Cloth
MFCD00673110

Specifications

Silver
Silver -colored gauze
Odorless
Ni:Cr; 80:20 wt%
Nickel Chromium, gauze
75 x 75mm
11106-97-1
Wire Cloth
MFCD00673110

Safety and Handling

SDS & Safety Documents

H351-H317: Suspected of causing cancer. May cause an allergic skin reaction.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit