Nickel Chromium gauze, Alfa Aesar™

Open area: 36%; Width of opening: 0.015 in.

Manufacturer: Alfa Aesar 40941HB

Quantity 300 x 300mm

 View more versions of this product

Catalog No. AA40941HB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11106-97-1
    Physical Form Wire Cloth
    Molecular Formula Ni:Cr; 80:20 wt%
    MDL Number MFCD00673110
    Chemical Name or Material Nickel Chromium, gauze

    Specifications

    Physical Form Wire Cloth
    Quantity 300 x 300mm
    Color Silver
    Odor Odorless
    Molecular Formula Ni:Cr; 80:20 wt%
    MDL Number MFCD00673110
    Appearance Silver -colored gauze
    CAS 11106-97-1
    Health Hazard 1 H351-H317: Suspected of causing cancer. May cause an allergic skin reaction.
    Chemical Name or Material Nickel Chromium, gauze

    H351-H317: Suspected of causing cancer. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit