Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Chromium Iron powder, -325 mesh, Thermo Scientific™

$750.17

Specifications

MDL Number MFCD01741927
Mesh Size −325 mesh
Chemical Name or Material Nickel Chromium Iron powder
Molecular Formula Ni:Cr:Fe; 58:27-31:7-11 wt%
Packaging Glass bottle
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA44888A1
View Documents Promotion Details
Thermo Scientific Chemicals
44888A1
1kg
Each for $750.17
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

MFCD01741927
Nickel Chromium Iron powder
Glass bottle
−325 mesh
Ni:Cr:Fe; 58:27-31:7-11 wt%
UN3089
Safety and Handling

Safety and Handling

P201-P202-P210-P240-P241-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H228-H317-H351

DOTInformation : Hazard Class: 4.1; Packaging Group: III

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit