Alfa Aesar™ Nickel Chromium Iron powder, -325 mesh

Manufacturer: Alfa Aesar™ 3660430

Quantity 250g

 View more versions of this product

Catalog No. AA3660430

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD01741927
    UN Number UN3089
    Chemical Name or Material Nickel Chromium Iron powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −325 Mesh
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H372-H351-H228-H319-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Flammable solid.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Quantity 250g
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD01741927
    UN Number UN3089
    Melting Point 1354° to 1413°C
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr:Fe; 72:14 to 17:6 to 10 wt%
    Chemical Name or Material Nickel Chromium Iron powder

    GHS H Statement
    H372-H351-H228-H319-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Flammable solid.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit