Learn More
Nickel Chromium Iron rod, 6.4mm (0.25 in.), hot finished, annealed, Thermo Scientific™
Specifications
Chemical Name or Material | Nickel Chromium Iron rod |
---|---|
Name Note | 6.4mm (0.25 in.), Hot finished, Annealed, Meets ASTM B166, Temper: soft |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cr:Fe; 72:14 to 17:6 to 10 wt% |
MDL Number | MFCD01741927 |
Weight | ≈269g/m |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA36455G1
|
Thermo Scientific Chemicals
36455G1 |
1 m | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
AA36455I7
|
Thermo Scientific Chemicals
36455I7 |
5 x 1 m | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nickel Chromium Iron rod | |
Ni:Cr:Fe; 72:14 to 17:6 to 10 wt% | |
≈269g/m |
6.4mm (0.25 in.), Hot finished, Annealed, Meets ASTM B166, Temper: soft | |
MFCD01741927 | |
Rod |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.