Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length, 99% (metals basis), Si nominal 1.5%

Click to view available options
Quantity:
5g
25g
100g
11106-97-1
CrNi
110.689
MFCD00673110
VNNRSPGTAMTISX-UHFFFAOYSA-N
nickel chromium,nickel-chromium,nicr,nickel chromium lump,chromium, compd. with nickel 1:1,chromium nickel,acmc-1byoe,nickel chromium powder,-140+325 mesh,nickel chromium wire, 1.0mm 0.04in dia,nickel chromium wire, 0.25mm 0.01in dia
11320920
chromium;nickel
[Cr].[Ni]
11106-97-1
CrNi
110.689
MFCD00673110
VNNRSPGTAMTISX-UHFFFAOYSA-N
nickel chromium,nickel-chromium,nicr,nickel chromium lump,chromium, compd. with nickel 1:1,chromium nickel,acmc-1byoe,nickel chromium powder,-140+325 mesh,nickel chromium wire, 1.0mm 0.04in dia,nickel chromium wire, 0.25mm 0.01in dia
11320920
chromium;nickel
[Cr].[Ni]

High resistivity material for electronics industry

Quantity 100g
Weight ≈0.21g/piece
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Concentration Si nomin.al 1.5%
Solubility Information Insoluble in water.
Odor Odorless
Physical Form ≈0.21g/piece
Percent Purity 99%
Chemical Name or Material Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length
Assay (metals basis)

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351

TSCA : Yes

Recommended Storage : Ambient temperatures